Learn More
(-)-Nicotine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 54-11-5 |
| Melting Point | -79°C |
| Density | 1.01 g/cm3 at 20°C |
| Boiling Point | 248.8°C/1013 hPa |
| MDL Number | MFCD00006369 |
| Quantity | 100 mL |
| Synonym | (-)-Nicotine, 3-(1-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-Nicotine, (-)-Nicotine |
| Molecular Weight (g/mol) | 162.23 |
| Grade | Research |