missing translation for 'onlineSavingsMsg'
Learn More

(-)-Nicotine Ditartrate, TRC

Product Code. 30041022
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041022 1 g 1g
30040446 100 mg 100mg
30007704 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041022 Supplier Toronto Research Chemicals Supplier No. TRCN4245981G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-31-6
Formula Weight 462.1486
InChI Formula InChI=1S/C10H14N2.2C4H6O6/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;2*5-1(3(7)8)2(6)4(9)10/h2,4,6,8,10H,3,5,7H2,1H3;2*1-2,5-6H,(H,7,8)(H,9,10)/t10-;2*1-,2-/m011/s1
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molecular Formula C10 H14 N2 . 2 C4 H6 O6
Molecular Weight (g/mol) 462.41
Percent Purity >95%
SMILES CN1CCC[C@H]1c2cccnc2.O[C@H]([C@@H](O)C(=O)O)C(=O)O.O[C@H]([C@@H](O)C(=O)O)C(=O)O
Synonym Nicotine ditartrate, 3-[(2S)-1-Methyl-2-pyrrolidinyl]pyridine (2R,3R)-2,3-dihydroxybutanedioate (1:2), Nicotine tartrate (1:2), (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:2), (-)-Nicotine ditartrate, (-)-Nicotine hydrogen (+)-tartrate, NSC 97238, Nicotine acid tartrate, Nicotine bitartrate, Nicotine hydrogen (+)-tartrate, Nicotine hydrogen tartrate, Nicotine tartrate, S-(-)-Nicotine hydrogen-(+)-tartrate
Chemical Name or Material Nicotine ditartrate
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.