missing translation for 'onlineSavingsMsg'
Learn More

β-Nicotinamide Adenine Dinucleotide, MP Biomedicals™

Product Code. 11427560 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
500 mg
5 g
10 g
Unit Size:
100mg
10g
500mg
5g
This item is not returnable. View return policy

Product Code. 11427560

Brand: MP Biomedicals 0210049910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acts as a major electron acceptor molecule in biological oxidations. β-Nicotinamide Adenine Dinucleotide, MP Biomedicals serves as a coenzyme of the dehydrogenases and acts as a hydrogen acceptor. β-NAD is one of the biologically active forms of

β-NAD occurs in living cells primarily in the oxidized state.

  • Serves as a coenzyme of hydride-transfer enzymes in dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols
  • Usually acts as a hydrogen acceptor, forming NADH, which then serves as a hydrogen donor in the respiratory chain
  • Used as a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins
  • Recently reported to be a P2Y11 agonist
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 53-84-9
Molecular Formula C21H27N7O14P2
Molecular Weight (g/mol) 663.43
MDL Number MFCD00150377
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
PubChem CID 15938971
ChEBI CHEBI:57540
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifikationer

Absorbance λ max (water): 259 ± 3nm
Melting Point 160°C (literature)
pH 2 to 4 (1% aq. soln.)
Quantity 10 g
Solubility Information Soluble in water (200mg/mL)
Formula Weight 663.4 (anhydrous)
Percent Purity ≥98%
Physical Form Powder
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.