accessibility menu, dialog, popup

UserName

CAS RN 53-84-9

IUPAC Name: 1-(5-{[({[5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl)-3-carbamoyl-1λ⁵-pyridin-1-ylium
Synonyms: nicotinamide adenine dinucleotide
Molecular Weight (g/mol): 663.43
Molecular Formula: C21H27N7O14P2
InChi Key: BAWFJGJZGIEFAR-UHFFFAOYNA-N
SMILES: NC(=O)C1=CC=C[N+](=C1)C1OC(COP([O-])(=O)OP(O)(=O)OCC2OC(C(O)C2O)N2C=NC3=C(N)N=CN=C23)C(O)C1O
Molecular Weight (g/mol) 
  • (3)
  • (26)
Quantity 
  • (10)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (8)
  • (1)
Physical Form 
  • (3)
  • (1)
  • (18)
Percent Purity 
  • (2)
  • (7)
  • (3)
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (12)
  • (1)
  • (5)
  • (3)
  • (1)
  • (3)
  • (3)
  • (26)
  • (10)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (8)
  • (1)
  • (3)
  • (1)
  • (18)
  • (2)
  • (7)
  • (3)
  • (3)
  • (2)
  • (2)