Learn More
Nicodicosapent, MedChemExpress
Product Code. 16467438
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467438 10 mg 10mg
16449917 5 mg 5mg
16413467 50 mg 50mg
16407416 100 mg 100mg
16400306 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16467438 Supplier Medchem Express Supplier No. HY17640
 Restricted Item
Estimated Shipment: 29-09-2026
MedChemExpress Nicodicosapent is a fatty acid niacin conjugate that is also an inhibitor of the sterol regulatory element binding protein (SREBP), a key regulator of cholesterol metabolism proteins such as PCSK9, HMG-CoA reductase, ATP citrate lyase, and NPC1L1.
Nicodicosapent is a fatty acid niacin conjugate that is also an inhibitor of the sterol regulatory element binding protein (SREBP), a key regulator of cholesterol metabolism proteins such as PCSK9, HMG-CoA reductase, ATP citrate lyase, and NPC1L1.
Specifications
Chemical Name or Material Nicodicosapent
CAS 1269181-69-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H39N3O2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (222.41 mM) ∣Ethanol : 2 mg/mL (4.45 mM; Need ultrasonic)
SMILES O=C(C1=CC=CN=C1)NCCNC(CCC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O
Molecular Weight (g/mol) 449.63
Formula Weight 449.63
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism