missing translation for 'onlineSavingsMsg'
Learn More

Nickel Acetate, MP Biomedicals

Product Code. 11448765 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448765 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448765 Supplier MP Biomedicals Supplier No. 0522479880

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6018-89-9
Quantity 100 g
Molecular Formula C4H14NiO8
MDL Number MFCD00066973
Synonym nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
InChI Key OINIXPNQKAZCRL-UHFFFAOYSA-L
SMILES O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
IUPAC Name nickel(2+) diacetate tetrahydrate
Molecular Weight (g/mol) 248.84
PubChem CID 62601
Formula Weight 176.8
Chemical Name or Material Nickel Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.