Learn More
Cytiva Ni Sepharose™ excel
Product Code. 13415187 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13415187 25 mL 25mL
12664557 100 mL 100mL
12621206 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13415187 Supplier Cytiva Supplier No. 17371201
Ni Sepharose excel affinity media for capture and purification of histidine-tagged proteins secreted into eukaryotic cell culture supernatants by immobilized metal ion affinity chromatography (IMAC).
  • Load eukaryotic cell culture samples containing secreted histidine-tagged proteins directly with retained binding capacity.
  • Increase target protein yield and decrease degradation through reduced and simplified sample handling.
  • Choose between several formats for screening and preparative purification of histidine-tagged proteins.
  • Ni Sepharose excel is an IMAC medium precharged with nickel ions very strongly bound to a chelating ligand, allowing loading of samples that usually cause stripping of metal ions.
  • Ni Sepharose excel is designed primarily for capture and purification of histidine-tagged proteins secreted into cell culture supernatants from eukaryotic cells such as insect cells or CHO cells.
TRUSTED_SUSTAINABILITY
Specifications
Buffer 0.01M HCl and 0.01M NaOH (one week), 10mM EDTA, 5mM DTT, 1M NaOH, 5mM TCEP, 20mM beta-Mercapto ethanol and 6M Guanidine-HCl (24 Hours), 500mM Imidazole and 100mM EDTA (2 Hours), 30% 2-Propanol
Product Type Ni Sepharose excel chromato graphy media
Quantity 25 mL
Flow Rate 150 to 600cm/hr.
pH Range 2 to 12
Particle Size 90μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4° to 30°C
For Use With (Application) Capture and purification of His-tagged Proteins secreted into eukaryotic cell culture supernatants by immobilized Metal ion affinity chromatography (IMAC)
Matrix Ni-Sepharose
compliance-icons
Signal Word
  • ||Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH208-Contains Nickel. May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.