Learn More
Cytiva NHS-Activated Sepharose™ 4 Fast Flow Affinity Media
Product Code. 10343240 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10343240 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10343240 Supplier Cytiva Supplier No. 17090601
 Restricted Item
In Stock
Use for coupling amino-containing smaller proteins and peptides. Cytiva Life Sciences™ NHS-Activated Sepharose™ 4 Fast Flow Affinity Media yields coupled products that are very stable, especially at high pH.
  • Composed of cross-linked 4% agarose beads, NHS activated for coupling amino-containing proteins and peptides
  • NHS (N-hydroxysuccinimide) coupling forms chemically stable amide bond with ligands containing primary amino groups
  • Resin provides spacer arm to coupled ligand
  • Fast Flow matrix gives high productivity
  • Coupled product is very stable, especially at high pH
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 mL
Stability Stable in commonly used aqueous Buffers provided that the ligand withstand the pH or the chemical environment
For Use With (Application) For Coupling Amino-Containing Smaller Proteins and Peptides
Storage Requirements 2°C to 8°C
Product Type NHS-Activated Sepharose 4 Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 3 to 13
Storage Buffer 100% Isopropanol
Ligand Type N-hydroxysuccinimide (NHS) Activated
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NHS-activated Sepharose 4 Fast Flow, 25 ml
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol