missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF90/ILF3 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00 EUR - 349.00 EUR
Specifications
| Antigen | NF90/ILF3 |
|---|---|
| Clone | 2H10G3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235103
|
Proteintech
68213-1-IG-150UL |
150 μL |
410.00 EUR 349.00 EUR / 150µL Save 61.00 EUR 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236068
|
Proteintech
68213-1-IG-20UL |
20 μL |
145.00 EUR 129.00 EUR / 20µL Save 16.00 EUR 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NF90/ILF3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NF90/ILF3. This antibody recognizes human, mouse, rat antigen. The NF90/ILF3 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| NF90/ILF3 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3609 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2H10G3 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| CBTF; DRBF; DRBP76; MMP4; MPHOSPH4; MPP4; NF-AT-90; NF110; NF90; NFAR; NFAR-1; NFAR2; TCP110; TCP80 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title