Learn More
Eppendorf™ Universal Platform for Model C76 Water Bath Shaker
Description
- Holds different combinations of flask clamps
- Maximum number of flasks: 82 (10mL), 16 (25mL), 14 (50 or 125mL), 8 (250mL), 6 (500mL), 4 (1L), or 2 (2L)
- Measures 16.5W × 12 in.L (42 × 30cm)
Specifications
Specifications
| For Use With (Equipment) | Model C76 water bath shaker |
| Type | Universal Platform |
| Dimensions (L x W) | 30.5 x 41.9 cm |