Learn More
Thermo Scientific™ Neutralization Solution for MagJET Plasmid DNA Kit
Product Code. 13535230 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
90 mL
Unit Size:
25mL
90mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13535230 25 mL 25mL
13525230 90 mL 90mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13535230 Supplier Thermo Scientific™ Supplier No. R2601
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures.
The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Specifications
Buffer Neutralization Solution
Product Line MagJET
Quantity 25 mL