missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NeuN Polyclonal Antibody

Product Code. 17896677
Change view
Click to view available options
Quantity:
100 μL
500 μL
Unit Size:
100µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17896677 100 μL 100µL
17826477 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17896677 Supplier Invitrogen™ Supplier No. PA5143567
  • ONLINE EXCLUSIVE PRICE
    500.00 EUR
    429.65 EUR / 100µL
    Save 70.35 EUR
    14% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken Polyclonal Antibody

NeuN Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IHC (PFA)

Fox-3, also known as NEUN, HRNBP3, RBFOX3 (RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuN
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RBFOX3
Gene Accession No. A6NFN3, Q8BIF2
Gene Alias D11Bwg0517e; FLJ56884; FLJ58356; fox 1 homolog (C. elegans) 3; fox1 homolog C; Fox-1 homolog C; FOX3; FOX-3; FOX3NeuN; hexaribonucleotide binding protein 3; hexaribonucleotide binding protein 3 (HRNBP3); Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding fox-1 homolog 3; RNA binding protein; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3
Gene Symbols RBFOX3
Host Species Chicken
Immunogen N-terminal 100 amino acids of human FOX3 expressed in and purified from E. coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146713, 287847, 52897
Target Species Human, Mouse, Rat, Monkey, Bovine
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.