Description
- Ultra-wraparound frame
- PC/TPR co-injected temples for greater comfort
- Panoramic field of vision
- Scratch and fog resistant coating
- Non-slip nose bridge
Specifications
Specifications
| Certifications/Compliance | CE, EN166 FT |
| Lens Type | Platinum™ |
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | CSP (Comfort Sensitivity Perception) |
| Product Type | Safety Glasses |
| No. per Pack | 1 Ea. |
| Weight (Metric) | 28 g |
| Lens Material | Polycarbonate |