missing translation for 'onlineSavingsMsg'
Learn More

Neramexane Hydrochloride, TRC

Product Code. 30475575
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475575 5 mg 5mg
30466420 2.5 mg 2.5mg
30464459 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475575 Supplier Toronto Research Chemicals Supplier No. TRCN3900935MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Neramexane Hydrochloride
CAS 209185-99-9
Molecular Formula C11H23N.HCl
Quantity 5 mg
Synonym 1,3,3,5,5-Pentamethylcyclohexanamine Hydrochloride, 1-Amino-1,3,3,5,5-pentamethylcyclohexane Hydrochloride
InChI Formula InChI=1S/C11H23N.ClH/c1-9(2)6-10(3,4)8-11(5,12)7-9;/h6-8,12H2,1-5H3;1H
SMILES Cl.NC1(C)CC(C)(C)CC(C)(C)C1
IUPAC Name 1,3,3,5,5-Pentamethylcyclohexan-1-amine Hydrochloride
Molecular Weight (g/mol) 205.77
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.