Learn More
Neostigmine Metilsulfate, Mikromol™
Product Code. 18041230
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18041230 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18041230 Supplier Mikromol Supplier No. MM0857.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Neostigmine Metilsulfate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Neostigmine Methyl Sulfate
CAS 51-60-5
Analyte or Component Names Neostigmine Methyl Sulfate
Shipping Condition Room Temperature
Molecular Formula C12 H19 N2 O2 . C H3 O4 S
Certifications/Compliance ISO 17034
Synonym Neostigmine metilsulfate, 3-[(Dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium methyl sulfate
InChI Formula InChI=1S/C12H19N2O2.CH4O4S/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;1-5-6(2,3)4/h6-9H,1-5H3;1H3,(H,2,3,4)/q+1;/p-1
SMILES COS(=O)(=O)[O-].CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C
IUPAC Name [3-(dimethylcarbamoyloxy)phenyl]-trimethylazanium;methyl sulfate
Molecular Weight (g/mol) 334.39
Quantity 250 mg
Formula Weight 334.1199 g/mol