Learn More
Neostigmine Metilsulfate, LoGiCal
Product Code. 30459885
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459885 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459885 Supplier LoGiCal Supplier No. LGCFOR0857.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Neostigmine Methyl Sulfate
CAS 51-60-5
Analyte or Component Names Neostigmine Methyl Sulfate
Shipping Condition Room Temperature
Molecular Formula C12 H19 N2 O2 . C H3 O4 S
Certifications/Compliance ISO 9001
Synonym Neostigmine metilsulfate, 3-[(Dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium methyl sulfate
InChI Formula InChI=1S/C12H19N2O2.CH4O4S/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;1-5-6(2,3)4/h6-9H,1-5H3;1H3,(H,2,3,4)/q+1;/p-1
Solution Type Neat
SMILES COS(=O)(=O)[O-].CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C
IUPAC Name [3-(dimethylcarbamoyloxy)phenyl]-trimethylazanium;methyl sulfate
Industry Type Clinical Testing
Molecular Weight (g/mol) 334.39
Quantity 10 mg
Formula Weight 334.1199 g/mol