Learn More
Invitrogen™ Neon™ Transfection System Pipette
Product Code. 10575614
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10575614 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10575614 Supplier Invitrogen™ Supplier No. MPP100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Neon™ Transfection System Pipette
The Neon™ Transfection System Pipette is designed specifically for use with the Neon™ Transfection System device (MPK5000). Each device (MPK5000) already comes with a pipette. The pipette is also sold separately for those labs needing and additional pipette or replacement for their existing pipette.
For more information on the product, please click here
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The Neon™ Transfection System Pipette comes with 1 x pipette. Refer to the Neon™ Transfection System users manual for protocol instructions. For use at room temperature under normal laboratory conditions.
For Use With Transfection Grade Nucleic Acid Purification Kits, siRNA Vectors, DNA Vectors
Product Line Neon
Type Electroporation Pipette Tip
For Use With (Equipment) Neon™ Transfection System
Quantity 1 each
Serum Compatible Yes
Shipping Condition Room Temperature
Starting Material Cell No. 2x10^4 - 6 x10^8 cells; Varies by cell type
System Type Neon™ Transfection System