missing translation for 'onlineSavingsMsg'
Learn More

Neohesperidin, 97.8%, MP Biomedicals™

Product Code. 11405011 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405011 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11405011 Supplier MP Biomedicals Supplier No. 0215581380
  • ONLINE EXCLUSIVE PRICE
    92.60 EUR
    88.20 EUR / 100mg
    Save 4.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Soluble in pyridine (20mg/mL-clear, yellow solution)

TRUSTED_SUSTAINABILITY

Specifications

CAS 13241-33-3
Color Tan
Molecular Formula C28H34O15
MDL Number MFCD16661259
Quantity 100 mg
Synonym hesperetin-7-o-neohesperidoside
InChI Key ARGKVCXINMKCAZ-CIDMPNHLSA-N
SMILES CC1C(C(C(C(O1)OC2C(C(C(OC2OC3=CC(=C4C(=O)CC(OC4=C3)C5=CC(=C(C=C5)OC)O)O)CO)O)O)O)O)O
IUPAC Name (2S)-7-[(2S,3R,4S,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3S,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-2,3-dihydrochromen-4-one
Molecular Weight (g/mol) 610.565
PubChem CID 134129340
Formula Weight 610.6
Physical Form Powder
Chemical Name or Material Neohesperidin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.