Learn More
Neodymium Boron Iron, REacton™
Product Code. 11316898
Click to view available options
Quantity:
50 g
250 g
2 x 500 g
Unit Size:
250g
500g
50g
Product Code. 11316898
Brand: Thermo Scientific Alfa Aesar 013642.30
Request Bulk or Custom Format
Nd:B:Fe; 15:8:77 atomic%
Neodymium Boron Iron applied to manufacture head actuators for computer hard disks, magnetic resonance imaging (MRI), magnetic guitar pickups, loudspeakers and headphones, mechanical e-cigarette firing switches and magnetic bearings and couplings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Neodymium Boron Iron
Name Note Magnet Alloy Additive
Color Gray-Silver
Physical Form Broken Lump
Quantity 250 g
Molecular Formula Nd:B:Fe; 15:8:77 atomic%
MDL Number MFCD02091729
Synonym Magnet Alloy Additive
Solubility Information Insoluble in water.
Odor Odorless
Appearance Silver gray broken lump