missing translation for 'onlineSavingsMsg'
Learn More

Nefopam hydrochloride, MedChemExpress

Product Code. p-200096918
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448658 10 mg 10mg
16444537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448658 Supplier Medchem Express Supplier No. HYB1057

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Nefopam hydrochloride (Fenazoxine hydrochloride) is a centrally-acting but non-opioid analgesic drug, for the relief of moderate to severe pain. Nefopam hydrochloride targets β-catenin protein level in mesenchymal cells in-vitro and in-vivo.

Nefopam hydrochloride (Fenazoxine hydrochloride) is a centrally-acting but non-opioid analgesic drug, for the relief of moderate to severe pain. Nefopam hydrochloride targets β-catenin protein level in mesenchymal cells in-vitro and in-vivo.

Specifications

Chemical Name or Material Nefopam hydrochloride
CAS 23327-57-3
Color Off-White
Purity Grade Notes Research
Quantity 10 mg
Synonym Fenazoxine hydrochloride
Solubility Information H2O : 12.5 mg/mL (43.13 mM; Need ultrasonic) ∣DMSO : 12.5 mg/mL (43.13 mM; Need ultrasonic)
SMILES CN1CCOC(C2=CC=CC=C2)C3=CC=CC=C3C1.[H]Cl
Molecular Weight (g/mol) 289.8
Percent Purity 99.78%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.