Learn More
Thermo Scientific™ Needles for Luer-LOK Priming Syringes
Product Code. 10035934 Shop All Thermo Scientific Products
Change view
Click to view available options
Needle Gauge:
22 G
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Unit Size
10035934 22 G Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10035934 Supplier Thermo Scientific™ Supplier No. 365RNL22
Estimated Shipment: 23-09-2026
Thermo Scientific™ Needles for Luer-LOK Priming Syringes are available for syringes with removable needles.
Thermo Scientific™ Needles for Luer-LOK Priming Syringes are available for syringes with removable needles.
TRUSTED_SUSTAINABILITY
Specifications
Description Needles for Luer-Lok Priming Syringes; 22 gauge; 2 in. Needle Length; 2/Pk.
Needle Gauge 22 G
Length (English) Needle 2 in.
Product Type Syringe Needle
Length (Metric) Needle 50.8 mm
For Use With (Equipment) LC Autosampler Syringe
Connection Type Removable