missing translation for 'onlineSavingsMsg'
Learn More

Nedocromil, MedChemExpress

Product Code. p-200101757
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485688 10 mg 10mg
16428387 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16485688 Supplier Medchem Express Supplier No. HY13448

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nedocromil suppresses the action or formation of multiple mediators, including histamine, leukotriene C4 (LTC4), and prostaglandin D2 (PGD2).

Nedocromil suppresses the action or formation of multiple mediators, including histamine, leukotriene C4 (LTC4), and prostaglandin D2 (PGD2).

Specifications

Chemical Name or Material Nedocromil
CAS 69049-73-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H17NO7
Quantity 10 mg
Synonym FPL 59002
Solubility Information DMSO : 16.67 mg/mL (44.89 mM; Need ultrasonic)
SMILES O=C(C1=CC(C2=CC3=C(N(CC)C(C(O)=O)=CC3=O)C(CCC)=C2O1)=O)O
Molecular Weight (g/mol) 371.34
Formula Weight 371.34
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.