missing translation for 'onlineSavingsMsg'
Learn More

NECAB3 Antibody, Novus Biologicals™

Product Code. 30228288 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30228288 20 μL 20µL
30230149 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30228288 Supplier Novus Biologicals Supplier No. NBP33556420ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NECAB3 Polyclonal antibody specifically detects NECAB3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NECAB3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:2000, ELISA
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias Amyloid beta A4 protein-binding family A member 2-binding protein, APBA2BPfamily A, member 2 binding protein, dJ63M2.4, dJ63M2.5, EFCBP3STIP3, EF-hand calcium binding protein 3, Neuronal calcium-binding protein 3, neuronal calcium-binding protein NECAB3, NIP1Nek2-interacting protein 1, N-terminal EF-hand calcium binding protein 3, N-terminal EF-hand calcium-binding protein 3, synaptotagmin interacting protein 2, synaptotagmin interacting protein STIP3, SYTIP2, XB51X11L-binding protein 51
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-362 of human NECAB3 (NP_112508.3).,, Sequence:, MACAGLLTVCLLRPPAPQPQPQTPRHPQLAPDPGPAGHTLFQDVFRRADKNDDGKLSFEEFQNYFADGVLSLGELQELFSGIDGHLTDNLETEKLCDYFSEHLGVYRPVLAALESLNRAVLAAMDATKLEYERASKVDQFVTRFLLRETVSQLQALQSSLEGASDTLEAQAHGWRSDAESVEAQSRLCGSRRAGRRALRSVSRSSTWSPGSSDTGRSSEAEMQWRLQVNRLQELIDQLECKAPRLEPLREEDLAKGPDLHILMAQRQVQVAEEGLQDFHRALRCYVDFTGAQSHCLHVSAQKMLDGASFTLYEFWQDEASWRRHQQSPGSKAFQRILIDHLRAPDTLTTVFFPASWWIMNNN
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 63941
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.