Learn More
Deutsch Neumann™ Rubber Safety Pipette Fillers
Product Code. 10313441
Change view
Click to view available options
Model:
Standard 1
Universal 2
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
10313441 Standard 1 Each
10418882 Universal 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10313441 Supplier Deutsch Neumann™ Supplier No. 4700001
  • ONLINE EXCLUSIVE PRICE
    5.60 EUR
    4.44 EUR / Pack of 1
    Save 1.16 EUR
    20% Off
Estimated Shipment: 03-12-2026
Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ Rubber Safety Pipette Fillers are manufactured from natural rubber.
  • Only two operating points
  • Excellent tensile strength and resilience
  • One-handed design is the simplest safety pipette available
Specifications
Color Red
Material Natural Rubber
Model Standard 1
Type Safety Pipette Filler
For Use With (Application) For comfortable pipetting toxic, corrosive, containing bacteria and malodorous substances
Quantity 1 Each