missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ Bis-Tris Mini Protein Gels, 3 to 12%, 1.0 mm

Product Code. p-200056759
Change view
Click to view available options
Wells:
15-well
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells unitSize
12126600 15-well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12126600 Supplier Invitrogen™ Supplier No. BN2012BX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Invitrogen NativePAGE Bis-Tris Gel System is a precast polyacrylamide mini-gel system that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations, and assessment of purity.

Invitrogen NativePAGE Bis-Tris Protein gels are precast polyacrylamide mini-gels that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations and assessment of purity. They are based on the blue native polyacrylamide gel electrophoresis (BN PAGE) technique developed by Schägger and von Jagow, which overcomes the limitations of traditional native electrophoresis by providing a near-neutral operating pH and detergent compatibility.

Invitrogen NativePAGE Bis-Tris Protein gels are precast polyacrylamide mini-gels that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations and assessment of purity. They are based on the blue native polyacrylamide gel electrophoresis (BN PAGE) technique developed by Schägger and von Jagow, which overcomes the limitations of traditional native electrophoresis by providing a near-neutral operating pH and detergent compatibility.

Features of the NativePAGE Bis-Tris Protein gels include:

  • Wide molecular weight resolving range of 15–10,000 kDa
  • Neutral-pH separation, which better preserves the native state of protein complexes
  • Resolution of all proteins regardless of their isoelectric point (pI)
  • Ability to analyze membrane-protein complexes in their native conformations
  • Ability to obtain better resolution than with traditional tris-glycine native electrophoresis

How NativePAGE Bis-Tris Protein gels work

In SDS-PAGE, SDS functions as a charge-shift molecule that denatures proteins by conferring on them a net negative charge and enables proteins to migrate towards the anode. BN PAGE uses Coomassie G-250 as the charge-shift molecule, which binds to proteins and confers a net negative charge while maintaining the proteins in their nondenatured, native state. The near-neutral pH of the NativePAGE Bis-Tris Protein gels provides maximum stability of both the proteins and the gel matrix, resulting in a highly sensitive method for analysis of native membrane-protein complexes and offering superior band resolution over traditional Tris-glycine gel systems.

For transferring proteins to a membrane, we recommend using NuPAGE transfer buffers for traditional wet transfer and PVDF membranes. NuPAGE transfer buffers maintain the neutral pH environment established during electrophoresis. Nitrocellulose membranes are not compatible for blotting since the membrane will bind the Coomassie G-250 dye very tightly. Alternatively, rapid semi-dry transfer can be done using the Power Blotter (Cat. No. PB0013) or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001) using PVDF membranes.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C. Do not freeze.
Gel Percentage 3 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Sample Loading Volume Up to 15 μL
Separation Range 30 to 10,000 kDa
Separation Type Native
Wells 15-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Wet Ice
Length (Metric) 8 cm
Width (Metric) 8 cm
Product Line NativePAGE
For Use With (Equipment) XCell SureLock Mini-Cell
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.