Learn More
Nateglinide, MedChemExpress
Product Code. 16464357
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464357 50 mg 50mg
16408488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16464357 Supplier Medchem Express Supplier No. HYB0422
 Restricted Item
Estimated Shipment: 30-11-2026
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
MedChemExpress Nateglinide, a D-phenylalanine derivative, is an orally active and short-acting insulinotropic agent and a DPP IV inhibitor. Nateglinide inhibits ATP-sensitive K+ channels in pancreatic β-cells. Nateglinide is used for the treatment of type 2 (non-insulin-dependent) diabetes mellitus.
Nateglinide, a D-phenylalanine derivative, is an orally active and short-acting insulinotropic agent and a DPP IV inhibitor. Nateglinide inhibits ATP-sensitive K+ channels in pancreatic β-cells. Nateglinide is used for the treatment of type 2 (non-insulin-dependent) diabetes mellitus.
Especificaciones
Chemical Name or Material Nateglinide
Physical Form Solid
CAS 105816-04-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H27NO3
For Use With (Application) Neuroscience-Neuromodulation
Synonym A4166 Senaglinide
Solubility Information DMSO : 100 mg/mL (315.04 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](CC1=CC=CC=C1)NC([C@H]2CC[C@H](C(C)C)CC2)=O
Molecular Weight (g/mol) 317.42
Formula Weight 317.42
Percent Purity 99.78%
Grade Research
Mostrar más Mostrar menos
Research purposes only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.