Learn More
Naproxen, Mikromol™
Product Code. 18040610
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18040610 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18040610 Supplier Mikromol Supplier No. MM0152.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Naproxen
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Naproxen
CAS 22204-53-1
Analyte or Component Names Naproxen
Shipping Condition Room Temperature
Molecular Formula C14 H14 O3
Certifications/Compliance ISO 17034
Synonym (+)-(S)-Naproxen, (+)-2-(6-Methoxy-2-naphthyl)propionic acid, (+)-Naproxen, (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, (S)-(+)-Naproxen, (S)-(+)-Naproxene, (S)-2-(6-Methoxy-2-naphthyl)propanoic acid, (S)-2-(6-Methoxy-2-naphthyl)propionic acid, (S)-Naproxen, Apo-Naproxen, Bonyl, CG 3117, Diocodal, Dysmenalgit, Equiproxen, Floginax, Laraflex, Laser, MNPA, Naixan, Napren, Naprium, Naprius, Naprosyn, Naprosyne, Naproxen, Naprux, Naxen, Nycopren, Panoxen, Prexan, Proxen, Proxine, RS 3540, Reuxen, Veradol, Xenar, d-2-(6-Methoxy-2-naphthyl)propionic acid, d-Naproxen, 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (alphaS)- (9CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (S)-, (+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid, (S)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid
InChI Formula InChI=1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m0/s1
SMILES COc1ccc2cc(ccc2c1)[C@H](C)C(=O)O
IUPAC Name (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
Molecular Weight (g/mol) 230.26
Quantity 500 mg
Formula Weight 230.0943 g/mol