Learn More
Naproxen, LoGiCal
Product Code. 30458166
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458166 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30458166 Supplier LoGiCal Supplier No. LGCFOR0152.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Naproxen
CAS 22204-53-1
Analyte or Component Names Naproxen
Shipping Condition Room Temperature
Molecular Formula C14 H14 O3
Certifications/Compliance ISO 9001
Synonym (+)-(S)-Naproxen, (+)-2-(6-Methoxy-2-naphthyl)propionic acid, (+)-Naproxen, (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, (S)-(+)-Naproxen, (S)-(+)-Naproxene, (S)-2-(6-Methoxy-2-naphthyl)propanoic acid, (S)-2-(6-Methoxy-2-naphthyl)propionic acid, (S)-Naproxen, Apo-Naproxen, Bonyl, CG 3117, Diocodal, Dysmenalgit, Equiproxen, Floginax, Laraflex, Laser, MNPA, Naixan, Napren, Naprium, Naprius, Naprosyn, Naprosyne, Naproxen, Naprux, Naxen, Nycopren, Panoxen, Prexan, Proxen, Proxine, RS 3540, Reuxen, Veradol, Xenar, d-2-(6-Methoxy-2-naphthyl)propionic acid, d-Naproxen, 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (alphaS)- (9CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (S)-, (+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid, (S)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid
InChI Formula InChI=1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m0/s1
Solution Type Neat
SMILES COc1ccc2cc(ccc2c1)[C@H](C)C(=O)O
IUPAC Name (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 230.26
Quantity 10 mg
Formula Weight 230.0943 g/mol