missing translation for 'onlineSavingsMsg'
Learn More

α-Naphthyl Phosphate Disodium Salt Hydrate , MP Biomedical

Product Code. 11462920 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462920 250 mg 250mg
11452920 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462920 Supplier MP Biomedicals Supplier No. 02100257.2
  • ONLINE EXCLUSIVE PRICE
    61.75 EUR
    58.80 EUR / 250mg
    Save 2.95 EUR
    4% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

α-Naphthyl Phosphate Disodium Salt Hydrate is a white, crystalline powder enzyme substrate used for cell biology and analysis.

  • Substrate for determination of phosphatase
  • Used as an organic chemical synthesis intermediate

Specifications

CAS 207569-06-0
Color White
Quantity 250 mg
Molecular Weight (g/mol) 268.12
Percent Purity ≥99%
Grade Not Available
Physical Form Crystalline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.