missing translation for 'onlineSavingsMsg'
Learn More

α-Naphthyl phosphate, 99.8%, MP Biomedicals™

Product Code. 11482920 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482920 5 g 5g
11472920 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482920 Supplier MP Biomedicals Supplier No. 0210025705
  • ONLINE EXCLUSIVE PRICE
    609.00 EUR
    579.60 EUR / 5g
    Save 29.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2183-17-7
Molecular Formula C10H7Na2O4P
Molecular Weight (g/mol) 268.12
MDL Number MFCD00041007
InChI Key QYURIFWAOPAPAJ-UHFFFAOYSA-L
Synonym disodium 1-naphthyl phosphate, sodium naphthalen-1-yl phosphate, disodium naphthyl phosphate, 1-naphthyl phsophoric acid sodium salt, disodium naphthalen-1-yl phosphate, alpha-naphthyl phosphate disodium salt, dipotassium naphthalen-1-yl phosphate, disodium 1-naphthyl phosphate hydrate, sodium alpha-naphthyl phosphate, 1-naphthyl disodium orthophosphate
PubChem CID 75132
IUPAC Name disodium naphthalen-1-yl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

Color White
Assay Percent Range 0.998
Quantity 5 g
Formula Weight 268.1
Physical Form Powder
Chemical Name or Material α-Naphthyl phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.