missing translation for 'onlineSavingsMsg'
Learn More

α-Naphthol,MP Biomedicals

Product Code. 11436695 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436695 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11436695 Supplier MP Biomedicals Supplier No. 0520418901

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 90-15-3
Molecular Formula C10H8O
Quantity 1 g
Synonym 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern
InChI Key KJCVRFUGPWSIIH-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2O
IUPAC Name naphthalen-1-ol
Molecular Weight (g/mol) 144.173
PubChem CID 7005
ChEBI CHEBI:10319
Formula Weight 144.18
Chemical Name or Material Alpha-Naphthol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.