missing translation for 'onlineSavingsMsg'
Learn More

Naphthol AS-LC, TRC

Product Code. 30137010
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137010 25 g 25g
30136515 10 g 10g
30003988 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137010 Supplier Toronto Research Chemicals Supplier No. TRCN23440025G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4273-92-1
Molecular Formula C19H16ClNO4
Molecular Weight (g/mol) 357.79
Synonym N-(4-Chloro-2,5-dimethoxyphenyl)-3-hydroxy-2-naphthalenecarboxamide, 2-Hydroxy-3-naphthoylamino-2',5'-dimethoxy-4-chloroanilide, 2-Hydroxynaphthalene-3-carboxylic Acid-2',5'-dimethoxy-4'-chlorophenylamidel, Anarthol AS-LC, C.I. 37555, C.I. Azoic Coupling Component 23, Conazoic BC, Dycosthol AS-LC, Kiwa Grounder LC, N-(2',5'-Dimethoxy-4'-chlorophenyl)-3-hydroxy-2-naphthalenecarboxamide, N-(4-Chloro-2,5-dimethoxyphenyl)-3-hydroxy-2-naphthamide, NSC 50688, NSC 73118, Naftol AS-LC, Naphtanilide LC, Naphtazol LC, Naphthol AS-LC, Naphthol LC, Naphtol AS-LC, Naphtol AS-LCLL, Sanatol LC;
IUPAC Name N-(4-chloro-2,5-dimethoxyphenyl)-3-hydroxynaphthalene-2-carboxamide
SMILES COc1cc(NC(=O)c2cc3ccccc3cc2O)c(OC)cc1Cl

Specifications

Quantity 25 g
InChI Formula InChI=1S/C19H16ClNO4/c1-24-17-10-15(18(25-2)9-14(17)20)21-19(23)13-7-11-5-3-4-6-12(11)8-16(13)22/h3-10,22H,1-2H3,(H,21,23)
Formula Weight 357.0768
Chemical Name or Material Naphthol AS-LC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.