Learn More
Naphthalene-2-sulfonamide, 97%, BLD Pharm™
Product Code. 30774400
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30774400 1 g 1g
30769124 25 g -
30767506 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30774400 Supplier BLD Pharm Supplier No. BD128851g
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
CAS 1576-47-2
Molecular Formula C10H9NO2S
MDL Number MFCD00051931
Quantity 1 g
SMILES O=S(C1=CC=C2C=CC=CC2=C1)(N)=O
Molecular Weight (g/mol) 207.25
Percent Purity 97%
Chemical Name or Material Naphthalene-2-sulfonamide