Learn More
Cytiva Nanosep™ Centrifugal Devices with Omega™ Membrane
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
300 kDa
Includes:
100/Pk.
24/Pk.
500/Pk.
Color:
Blue
Clear
Gray
Orange
Red
Unit Size:
Pack of 100
Pack of 24
Pack of 500
Description
- Simple, reliable,concentrating and desalting of 50 to 500 μL samples
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
| Color | Blue |
| Description | EFA: 0.3 cm2 |
| Material (Membrane) | PES |
| MWCO | 10 kDa |
| Sterility | Non-sterile |
| Housing Material | Polypropylene |
| Includes | 500/Pk. |
| Volume (Metric) | 50 to 500 μL (Sample) |