Description
Certified standards for verifying instrument performance to factory specifications are available separately.
For use with Thermo Scientific NanoDrop Spectrophotometers.
- Ten times more concentrated compared to other K2Cr2O7 calibration solutions
- Packaged in 0.5mL snap-top glass ampules
- Intended for single use only
Specifications
Specifications
| Type | CF-1 Calibration Check Fluid |
| For Use With (Equipment) | NanoDrop 8000, NanoDrop Lite, NanoDrop 2000, NanoDrop 2000c spectrophotometers |
| Packaging | 0.5 mL snap-top glass ampules |