Learn More
Thermo Scientific™ NanoDrop™ CF-8 Calibration Check Kit
Thermo Scientific™ NanoDrop CF-8 Calibration Check Kit is an aqueous potassium dichromate (K2Cr2O7) solution for use in confirming spectrophotometer calibration.
Brand: Thermo Scientific™ CHEM-CF-8
Description
Thermo Scientific™ NanoDrop CF-8 Calibration Check Kit is an aqueous potassium dichromate (K2Cr2O7) solution for use in confirming spectrophotometer calibration.
For use with the Thermo Scientific NanoDrop 8000 Spectrophotometer.- Ten times more concentrated than other commercially available K2Cr2O7 solutions
- Packaged in 0.5mL snap-top glass ampules
- Single-use
Includes:
(2) CF-1 Ampules and 8-well PCR tube strip.