Learn More
Thermo Scientific™ NanoDrop™ CF-8 Calibration Check Kit
Thermo Scientific™ NanoDrop CF-8 Calibration Check Kit is an aqueous potassium dichromate (K2Cr2O7) solution for use in confirming spectrophotometer calibration.
Brand:  Thermo Scientific™ CHEM-CF-8
Product Code. 16411591
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Thermo Scientific™ NanoDrop CF-8 Calibration Check Kit is an aqueous potassium dichromate (K2Cr2O7) solution for use in confirming spectrophotometer calibration.
For use with the Thermo Scientific NanoDrop 8000 Spectrophotometer.
  • Ten times more concentrated than other commercially available K2Cr2O7 solutions
  • Packaged in 0.5mL snap-top glass ampules
  • Single-use
Includes:
(2) CF-1 Ampules and 8-well PCR tube strip.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions