Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 555, Chromotek
Description
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Specifications
Specifications
| Antigen | Nano-Secondary™ |
| Conjugate | CoraLite Plus 555 |
| Host Species | Alpaca |
| Quantity | 100 μL |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Form | Liquid |
| Isotype | IgG |