Learn More
NAlpha, NEpsilon-Bis-boc-L-lysine, TRC
Product Code. 30240892
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240892 1 g 1g
30240579 5 g 5g
30240533 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30240892 Supplier Toronto Research Chemicals Supplier No. TRCB6569951G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 2483-46-7
Molecular Formula C16 H30 N2 O6
Molecular Weight (g/mol) 346.42
Synonym L-Lysine, N2,N6-bis[(1,1-dimethylethoxy)carbonyl]-, Lysine, N2,N6-dicarboxy-, N2,N6-di-tert-butyl ester (7CI), Lysine, N2,N6-dicarboxy-, N2,N6-di-tert-butyl ester, L- (8CI), N2,N6-Bis[(1,1-dimethylethoxy)carbonyl]-L-lysine, (S)-2,6-Bis(tert-butoxycarbonylamino)hexanoic acid, (S)-2,6-Bis(tert-butyloxycarbonylamino)hexanoic acid, Di(N-Boc)-L-lysine, Di-tert-butoxycarbonyl-L-lysine, Di[N-(tert-Butoxycarbonyl)]-L-lysine, N,N'-Bis(tert-butoxycarbonyl)-(S)-lysine, N,N'-Bis-tert-butoxycarbonyl-L-lysine, N,N'-Di-tert-butoxycarbonyl-L-lysine, N2,N6-Bis(tert-butoxycarbonyl)-L-lysine, NSC 164052
IUPAC Name (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)O
Specifica
InChI Formula InChI=1S/C16H30N2O6/c1-15(2,3)23-13(21)17-10-8-7-9-11(12(19)20)18-14(22)24-16(4,5)6/h11H,7-10H2,1-6H3,(H,17,21)(H,18,22)(H,19,20)/t11-/m0/s1
Quantity 1 g
Formula Weight 346.2104
Shipping Condition Room Temperature
Chemical Name or Material Na, Ne-Bis-boc-L-lysine
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.