missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Ngamma-xanthyl-L-asparagine, 98%, Thermo Scientific™

Product Code. 15420519
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420519 25 g 25g
15410519 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420519 Supplier Thermo Scientific Alfa Aesar Supplier No. H63690.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65420-40-8
Molecular Formula C22H24N2O6
Molecular Weight (g/mol) 412.44
MDL Number MFCD00066157
InChI Key YMGDQLXBNMRJMR-UHFFFAOYNA-N
Synonym boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
PubChem CID 7017974
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 177°C to 181°C (decomposition)
Color Orange
Beilstein 5172403
Quantity 25 g
Formula Weight 412.44
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nalpha-Boc-Ngamma-xanthyl-L-asparagine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.