Learn More
Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Product Code. 13477649 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
1 and 1.5 mL Vials
2 mL Vials
5 mL Vials
Vial Capacity:
25
81
100
Quantity:
1/Cs.
10/Cs.
20/Cs.
40/Cs.
80/Cs.
Unit Size:
Pack of 10
Pack of 20
Pack of 25
Pack of 40
Pack of 81
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Vial Capacity Quantity Unit Size
13477649 5 mL Vials 81 1/Cs. Pack of 81
12874372 1 and 1.5 mL Vials 100 10/Cs. Pack of 10
12745918 2 mL Vials 25 80/Cs. Pack of 25
13497649 2 mL Vials 81 40/Cs. Pack of 40
13487649 2 mL Vials 81 20/Cs. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13477649 Supplier Thermo Scientific™ Supplier No. CS509X10
  • ONLINE EXCLUSIVE PRICE
    66.89 EUR
    53.19 EUR / Pack of 81
    Save 13.70 EUR
    20% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Obtain a comprehensive solution when plastic is preferred to fiberboard.
Obtain a comprehensive solution when plastic is preferred to fiberboard. Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems fit in Thermo Scientific stainless-steel, ultra-low temperature freezer racks.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Type Cryogenic Storage Box
Capacity (Metric) 5 mL
For Use With (Equipment) Locator Jr. Plus, Locator 4 Plus, Locator 6 Plus
Height (English) Exterior 4 in.
Vial Capacity 81
Holds 5 mL Vials
Quantity 1/Cs.