missing translation for 'onlineSavingsMsg'
Learn More

Naled 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312103
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312103 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312103 Supplier Dr. Ehrenstorfer Supplier No. DRES15410000AL1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naled
CAS 300-76-5
Analyte or Component Names Naled
Shipping Condition Room Temperature
Molecular Formula C4 H7 Br2 Cl2 O4 P
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C4H7Br2Cl2O4P/c1-10-13(9,11-2)12-3(5)4(6,7)8/h3H,1-2H3
SMILES COP(=O)(OC)OC(Br)C(Cl)(Cl)Br
IUPAC Name (1,2-dibromo-2,2-dichloroethyl) dimethyl phosphate
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Molecular Weight (g/mol) 380.78
Quantity 5 x 1 mL
Formula Weight 377.7826 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.