missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAGA Polyclonal Antibody
GREENER_CHOICE

Product Code. 15749390 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15749390 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15749390 Supplier Invitrogen™ Supplier No. PA551433

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: WLAWERFRCN INCDEDPKNC ISEQLFMEMA DRMAQDGWRD MGYTYLNIDD CWIGGRDASG RLMPDPKRFP HGIPFLADYV HSLGLKLGIY ADMGNFTCMG YPGTTLDKVV QDAQTFAEWK VDMLKLDGCF ST Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

NAGA (N-acetylgalactosaminidase, alpha), also known as alpha-galactosidase B or GALB, is a 411 lysosomal protein belonging to the glycosyl hydrolase 27 family that may exist as a homodimer and plays a critical role in glycolipid breakdown. NAGA encodes alpha-N-acetylgalactosaminidase, a lysosomal enzyme, which cleaves alpha-N-acetylgalactosaminyl groups from glycoconjugates. Mapping to human chromosome 22q13. 2, NAGA defects are the cause of an autosomal recessive disorder with three phenotypes, known as Schindler disease (types I, II and III) or NAGA deficiency (types I, II and III). Characterized by neurologic manifestations that range in severity, Schindler disease type I is the most severe form, followed by type III, which may have mild-to-moderate effects. Schindler disease type II, also known as Kanzaki disease, is characterized by mild intellectual impairment and angiokeratoma corporis diffusum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAGA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAGA
Gene Accession No. P17050
Gene Alias Acetylgalactosaminidase, alpha-N- (alpha-galactosidase B); alpha-galactosidase B; Alpha-N-acetylgalactosaminidase; D22S674; EC 3.2.1; EC 3.2.1.49; galactosidasealphaB; galactosidase-alphaB; GALB; N-acetyl galactosaminidase, alpha; N-acetylgalactosaminidase, alpha-; Naga
Gene Symbols NAGA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NAGA. Recombinant protein control fragment (Product #RP-91478).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4668
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.