missing translation for 'onlineSavingsMsg'
Learn More

Nafion, TRC

Product Code. 17942665
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942665 5 g 5g
30136747 25 g 25g
17914965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17942665 Supplier Toronto Research Chemicals Supplier No. TRCN2150035G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31175-20-9
Molecular Formula (C2F4)x • (C2F3)y • (OC3F6)z • C2HF4O4S • x(H2O)
Synonym 1,1,2,2-Tetrafluoro-2-[1,2,2-trifluoro-1-(trifluoromethyl)-2-[(1,2,2-trifluorovinyl)oxy]ethoxy]-ethanesulfonic Acid Polymer with Tetrafluoroethylene, 2-[1-[Difluoro[(trifluoroethenyl)oxy]methyl]-1,2,2,2-tetrafluoroethoxy]-1,1,2,2-tetrafluoro-ethanesulfonic Acid Polymer with Tetrafluoroethene, Tetrafluoroethene Polymer with 2-[1-[Difluoro[(trifluoroethenyl)oxy]methyl]-1,2,2,2-tetrafluoroethoxy]-1,1,2,2-tetrafluoroethanesulfonic Acid, Flemion R Nafion 663492 PSA 1 PSA 2, Perfluoro-3,6-dioxa-4-methyl-7-octene-1-sulfonic acid-tetrafluoroethylene Copolymer, Perfluoro-3,6-dioxa-4-methyl-7-octenesulfonic acid-tetrafluoroethylene Copolymer, Perfluoro-3,6-dioxa-4-methyl-7-octenesulfonic acid-tetrafluoroethylene Polymer, Perfluoro[2-(2-sulfonylfluoroethoxy)propyl vinyl ether]-tetrafluoroethylene Copolymer, SHF 301, SPL 2, SPL 5, Nafion (NR50) Tetrafluoroethylene-perfluoro-3,6-dioxa-4-methyl-7-octenesulfonic Acid Copolymer;
IUPAC Name 1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(2,2,3,4,4,5,5-heptafluorohexan-3-yloxy)propan-2-yl]oxyethanesulfonic acid
SMILES OS(C(F)(F)C(F)(F)OC(F)(C(F)(F)F)C(F)(F)OC(C(F)(C(F)(F)C)F)(F)C(F)(F)C)(=O)=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C11H7F17O5S/c1-3(12,13)5(16,17)6(18,4(2,14)15)32-9(23,24)7(19,8(20,21)22)33-10(25,26)11(27,28)34(29,30)31/h1-2H3,(H,29,30,31)
Formula Weight 573.9743
Chemical Name or Material Nafion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.