Learn More
NAD+, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 53-84-9 |
| Molecular Formula | C21H27N7O14P2 |
| MDL Number | MFCD00036253 |
| Quantity | 25 g |
| SMILES | O[C@H]1[C@H](N2C3=C(N=C2)C(N)=NC=N3)O[C@@H]([C@H]1O)COP(OP(OC[C@H]4O[C@H]([C@@H]([C@@H]4O)O)[N+]5=CC=CC(C(N)=O)=C5)([O-])=O)(O)=O |
| Molecular Weight (g/mol) | 663.43 |
| Percent Purity | 98% |
| Chemical Name or Material | NAD+ |