missing translation for 'onlineSavingsMsg'
Learn More

Nabumetone, MedChemExpress

Product Code. 16470998
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470998 5 g 5g
16402239 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16470998 Supplier Medchem Express Supplier No. HYB0559
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nabumetone is an orally active non-acidic anti-inflammatory agent, acts as a potent and selective COX-2 inhibitor, and is the prodrug of the active metabolite 6MNA.

Nabumetone is an orally active non-acidic anti-inflammatory agent, acts as a potent and selective COX-2 inhibitor, and is the prodrug of the active metabolite 6MNA.

Specifications

Chemical Name or Material Nabumetone
CAS 42924-53-8
Color White
Purity Grade Notes Research
Molecular Formula C15H16O2
Quantity 5 g
Synonym BRL14777
Solubility Information DMSO : ≥ 100 mg/mL (438.04 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(CCC1=CC=C2C=C(OC)C=CC2=C1)=O
Molecular Weight (g/mol) 228.29
Formula Weight 228.29
Percent Purity 99.63%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.