missing translation for 'onlineSavingsMsg'
Learn More

N8-Acetylspermidine dihydrochloride, MedChemExpress

Product Code. 16467497
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467497 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16467497 Supplier Medchem Express Supplier No. HY113253A
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress N8-Acetylspermidine dihydrochloride is a polyamine.

N8-Acetylspermidine dihydrochloride is a polyamine.

Specifications

Chemical Name or Material N8-Acetylspermidine dihydrochloride
Physical Form Solid
CAS 34450-15-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C9H23Cl2N3O
Solubility Information DMSO : 100 mg/mL (384.32 mM; Need ultrasonic) ∣H2O : 50 mg/mL (192.16 mM; Need ultrasonic)
SMILES CC(NCCCCNCCCN)=O.Cl.Cl
Molecular Weight (g/mol) 260.2
Formula Weight 260.2
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.