missing translation for 'onlineSavingsMsg'
Learn More

N6-methyladenosine (m6A) Antibody, Novus Biologicals™

Product Code. 30232175 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30232175 20 μL 20µL
30230665 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30232175 Supplier Novus Biologicals Supplier No. NBP33319420ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Nonspecies specific samples. It is validated for ELISA,Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen N6-methyladenosine (m6A)
Applications ELISA, Dot Blot
Classification Monoclonal
Conjugate Unconjugated
Dilution ELISA Recommended starting concentration is 1 μg/mL, Dot Blot 1:500 - 1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine
Host Species Mouse
Immunogen Chemical compounds corresponding to N6-methyladenosine (m6A).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.