missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Nonspecies specific samples. It is validated for ELISA,Dot Blot
Specifications
Specifications
| Antigen | N6-methyladenosine (m6A) |
| Applications | ELISA, Dot Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Dilution | ELISA Recommended starting concentration is 1 μg/mL, Dot Blot 1:500 - 1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine |
| Host Species | Mouse |
| Immunogen | Chemical compounds corresponding to N6-methyladenosine (m6A). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?