Learn More
N4-Benzoyl-2’-deoxy-5’-O-DMT-cytidine 3’-CE Phosphoramidite, TRC
Specifications
Specifications
| Chemical Name or Material | N4-Benzoyl-2’-deoxy-5’-O-DMT-cytidine 3’-CE Phosphoramidite |
| CAS | 102212-98-6 |
| Molecular Formula | C46H52N5O8P |
| Quantity | 1 g |
| Synonym | N-Benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-2'-deoxy-cytidine 3'-[2-cyanoethyl bis(1-methylethyl)phosphoramidite], N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine-3'-O-[O-(2-cyanoethyl)-N,N'-diisopropylphosphoramidite], DMT-dC(bz) Phosphoramidite |
| InChI Formula | InChI=1S/C46H52N5O8P/c1-32(2)51(33(3)4)60(57-29-13-27-47)59-40-30-43(50-28-26-42(49-45(50)53)48-44(52)34-14-9-7-10-15-34)58-41(40)31-56-46(35-16-11-8-12-17-35,36-18-22-38(54-5)23-19-36)37-20-24-39(55-6)25-21-37/h7-12,14-26,28,32-33,40-41,43H,13,29-31H2,1-6H3,(H,48,49,52,53)/t40-,41+,43+,60?/m0/s1 |
| SMILES | O=C1N(C=CC(NC(C2=CC=CC=C2)=O)=N1)[C@@H]3O[C@@H]([C@H](C3)OP(OCCC#N)N(C(C)C)C(C)C)COC(C4=CC=C(OC)C=C4)(C5=CC=CC=C5)C6=CC=C(OC)C=C6 |
| IUPAC Name | (2R,3S,5R)-5-(4-benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite |
| Molecular Weight (g/mol) | 833.91 |
| Formula Weight | 833.36 |