missing translation for 'onlineSavingsMsg'
Learn More

N2'-Deacetyl-N2'-[3-(methylthio)-1-oxopropyl]-maytansine-d3, TRC

Product Code. 30356658
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356658 5 mg 5mg
30351698 2.5 mg 2.5mg
30359231 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30356658 Supplier Toronto Research Chemicals Supplier No. TRCD1990375MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N2'-Deacetyl-N2'-[3-(methylthio)-1-oxopropyl]-maytansine-d3
Molecular Formula C36H47D3ClN3O10S
Quantity 5 mg
Synonym S-Me-DM1-d3
InChI Formula InChI=1S/C36H50ClN3O10S/c1-20-11-10-12-27(47-8)36(45)19-26(48-34(44)38-36)21(2)32-35(4,50-32)28(49-33(43)22(3)39(5)29(41)13-14-51-9)18-30(42)40(6)24-16-23(15-20)17-25(46-7)31(24)37/h10-12,16-17,21-22,26-28,32,45H,13-15,18-19H2,1-9H3,(H,38,44)/b12-10+,20-11+/t21-,22+,26+,27-,28+,32+,35+,36+/m1/s1/i8D3
SMILES C[C@]12[C@@]([C@@H]([C@]3([H])C[C@]([C@@H](/C=C/C=C(CC4=CC(N(C(C[C@@H]2OC([C@H](C)N(C)C(CCSC)=O)=O)=O)C)=C(C(OC)=C4)Cl)\C)OC([2H])([2H])[2H])(NC(O3)=O)O)C)([H])O1
IUPAC Name (14S,16S,32S,33S,2R,4S,10E,12E,14R)-86-chloro-14-hydroxy-85-methoxy-14-(methoxy-d3)-33,2,7,10-tetramethyl-12,6-dioxo-7-aza-1(6,4)-oxazinana-3(2,3)-oxirana-8(1,3)-benzenacyclotetradecaphane-10,12-dien-4-yl N-methyl-N-(3-(methylthio)propanoyl)-L-alaninate
Molecular Weight (g/mol) 755.33
Formula Weight 754.3094
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.